Skip to Content
Merck
All Photos(1)

Documents

N5396

Supelco

19-Nortestosterone 17-dodecanoate

Synonym(s):

Laurabolin, Nandrolone laurate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
EC Number:
UNSPSC Code:
41116107

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

InChI

1S/C30H48O3/c1-3-4-5-6-7-8-9-10-11-12-29(32)33-28-18-17-27-26-15-13-22-21-23(31)14-16-24(22)25(26)19-20-30(27,28)2/h21,24-28H,3-20H2,1-2H3/t24-,25+,26+,27-,28-,30-/m0/s1

InChI key

OXXNTXVXBWLYQE-PVHICTMWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mickael Doué et al.
Drug testing and analysis, 7(7), 603-608 (2014-09-30)
High throughput screening is essential for doping, forensic, and food safety laboratories. While hyphenated chromatography-mass spectrometry (MS) remains the approach of choice, recent ambient MS techniques, such as direct analysis in real time (DART), offer more rapid and more versatile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service