Skip to Content
Merck
All Photos(3)

Documents

HPA021818

Sigma-Aldrich

Anti-MPP3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Discs large homolog 3, Anti-MAGUK p55 subfamily member 3, Anti-Protein MPP3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VCLVDVEPEALKQLRTSEFKPYIIFVKPAIQEKRKTPPMSPACEDTAAPFDEQQQEMAASAAFIDRHYGHLVDAVLVKEDLQGAYSQLKVVL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MPP3(4356)

General description

MPP3 (Membrane protein, palmitoylated 3, MAGUK p55 subfamily member 3) is a cytoplasmic protein belonging to the MAGUK family molecules. It is expressed in various tissues including normal lung cells. MPP3 is composed of a pair of Lin2/Lin7-binding (L27) domains, a PDZ domain, a Src-homology 3 (SH3) domain and a guanylate kinase homologous (GuK) domain. It exhibits high homology with Dlg in Drosophila.

Immunogen

MAGUK p55 subfamily member 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MPP3 (Membrane protein, palmitoylated 3, MAGUK p55 subfamily member 3) plays an important role in the maintenance of epithelial structure through cell-cell interactions. The N-terminal domain of MPP3 binds directly to the cytoplasmic domain of CADM1 (Cell adhesion molecule 1) and forms a complex, which further connects it to the PI3K (phosphoinositide 3-kinase) for its activation. The activation of PI3K is highly required to identify the actin cytoskeleton and form epithelial cell structure. A study reports that MPP3 in conjugation with TSLC1 (tumor suppressor in lung cancer 1) shows tumor suppressive property in human lung cancer cells and any change in this interactions might lead to malignancy and tumor growth.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75728

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigefumi Murakami et al.
PloS one, 9(2), e82894-e82894 (2014-02-08)
CADM1 (Cell adhesion molecule 1), a cell adhesion molecule belonging to the immunoglobulin superfamily, is involved in cell-cell interaction and the formation and maintenance of epithelial structure. Expression of CADM1 is frequently down-regulated in various tumors derived from epithelial cells.
Hiroshi Fukuhara et al.
Oncogene, 22(40), 6160-6165 (2003-09-19)
We have previously identified the tumor suppressor in lung cancer 1 (TSLC1) gene as a novel tumor suppressor in human non-small cell lung cancer (NSCLC) by functional complementation. TSLC1 encodes a membrane glycoprotein belonging to an immunoglobulin superfamily and participates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service