Skip to Content
Merck
All Photos(1)

Key Documents

EMU011661

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Icam1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€194.00
50 μG
€343.00

€194.00


Estimated to ship on21 April 2025



Select a Size

Change View
20 μG
€194.00
50 μG
€343.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€194.00


Estimated to ship on21 April 2025


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGTACATACGTGTGCCATGCCTTTAGCTCCCATGGGAATGTCACCAGGAATGTGTACCTGACAGTACTGTACCACTCTCAAAATAACTGGACTATAATCATTCTGGTGCCAGTACTGCTGGTCATTGTGGGCCTCGTGATGGCAGCCTCTTATGTTTATAACCGCCAGAGAAAGATCAGGATATACAAGTTACAGAAGGCTCAGGAGGAGGCCATAAAACTCAAGGGACAAGCCCCACCTCCCTGAGCCTGCTGGATGAGACTCCTGCCTGGACCCCCTGCAGGGCAACAGCTGCTGCTGCTTTTGAACAGAATGGTAGACAGCATTTACCCTCAGCCACTTCCTCTGGCTGTCACAGAACAGGATGGTGGCCTGGGGGATGCACACTTGTAGCCTCAGAGCTAAGAGGACTCGGTGGATGG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weibao Xiao et al.
Experimental eye research, 138, 145-152 (2015-07-19)
FTY720 is a promising drug in attenuating multiple sclerosis, prolonging survival of organ allograft, and many other protective effects. Its mechanism of action is considered to be mediated by the internalization of sphingosine 1-phosphate receptors (S1PRs). In the current study
Chunhua Jin et al.
Molecular medicine (Cambridge, Mass.), 20, 280-289 (2014-06-12)
The myocardial inflammatory response contributes to cardiac functional injury associated with heart surgery obligating global ischemia/reperfusion (I/R). Toll-like receptors (TLRs) play an important role in the mechanism underlying myocardial I/R injury. The aim of this study was to examine the
Fumitake Ito et al.
The Journal of clinical endocrinology and metabolism, 99(6), 2188-2197 (2014-03-13)
Monocyte adhesion to endothelial cells is an important initial event in atherosclerosis and is partially mediated by adhesion molecule expression on the cell surface. Although estrogens inhibit atherosclerosis development, effects of coadministered progestogen remain controversial. We examined the effects of
Nicolas J Pillon et al.
American journal of physiology. Endocrinology and metabolism, 309(1), E35-E44 (2015-05-07)
Obesity is associated with inflammation and immune cell recruitment to adipose tissue, muscle and intima of atherosclerotic blood vessels. Obesity and hyperlipidemia are also associated with tissue insulin resistance and can compromise insulin delivery to muscle. The muscle/fat microvascular endothelium
Shiro Koizume et al.
Molecular cancer, 14, 77-77 (2015-04-17)
Elucidation of the molecular mechanisms by which cancer cells overcome hypoxia is potentially important for targeted therapy. Complexation of hypoxia-inducible factors (HIFs) with aryl hydrocarbon receptor nuclear translocators can enhance gene expression and initiate cellular responses to hypoxia. However, multiple

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service