Skip to Content
Merck
All Photos(2)

Key Documents

E0904

Sigma-Aldrich

pBICEP-CMV-2 Expression Vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

shipped in

dry ice

storage temp.

−20°C

Application

For stable, cytoplasmic expression of N-terminal Met-3xFLAG fusion proteins from bicistronic mRNA. Stable expression is accomplished by neomycin selection (G 418 sulfate). Supplied with a pBICEP-CMV-2-lacZ Control Vector.

Other Notes

Browse additional application references in our FLAG® Literature portal.

Legal Information

3xFLAG is a trademark of Sigma-Aldrich Co. LLC
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
pBICEP-CMV is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service