Skip to Content
Merck
All Photos(1)

Key Documents

56211

Sigma-Aldrich

LAPAO

≥95% (HPLC)

Synonym(s):

3-Dodecylamido-N,N′-dimethylpropyl amine oxide, 3-Lauramidopropyl-N,N-dimethylamine oxide, N-[3-(Dimethylamino)propyl]dodecanamide N-oxide, Dodecylamidopropyldimethylamine oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H36N2O2
CAS Number:
Molecular Weight:
300.48
Beilstein:
2118734
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (HPLC)

mol wt

300.48 g/mol

technique(s)

electrophoresis: suitable

impurities

insoluble matter, passes filter test

λ

1 % (w/v) in H2O

UV absorption

λ: 260 nm Amax: ≤0.20
λ: 280 nm Amax: ≤0.15

SMILES string

CCCCCCCCCCCC(=O)NCCC[N+](C)(C)[O-]

InChI

1S/C17H36N2O2/c1-4-5-6-7-8-9-10-11-12-14-17(20)18-15-13-16-19(2,3)21/h4-16H2,1-3H3,(H,18,20)

InChI key

JNGWKQJZIUZUPR-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M R Block et al.
Biochemistry, 25(2), 374-379 (1986-01-28)
The mitochondrial adenine nucleotide (AdN) carrier can assume two conformational states that are trapped by the specific inhibitors of AdN transport carboxyatractyloside (CATR) and bongkrekic acid (BA). When the AdN carrier protein was extracted from beef heart mitochondria by the
Cécile Dahout-Gonzalez et al.
Acta crystallographica. Section D, Biological crystallography, 59(Pt 12), 2353-2355 (2003-12-04)
The ADP/ATP carrier is an integral membrane protein located in the mitochondrial inner membrane. It mediates the exchange of cytosolic ADP for ATP generated in the mitochondrial matrix. Here, the purification and crystallization of the bovine ADP/ATP carrier in complex
Sebastian F Zenk et al.
Microbiology (Reading, England), 153(Pt 8), 2405-2415 (2007-07-31)
Type III secretion systems (T3SSs or secretons) are central virulence factors of many Gram-negative bacteria, used to inject protein effectors of virulence into eukaryotic host cells. Their overall morphology, consisting of a cytoplasmic region, an inner- and outer-membrane section and
Claire Ramus et al.
Molecular & cellular proteomics : MCP, 5(1), 68-78 (2005-10-12)
The work presented here focuses on the development of a method adapting isotope labeling of proteins with ICAT to the study of highly hydrophobic proteins. Conditions for the labeling of proteins were first established using two standard soluble proteins and
Marjolaine Noirclerc-Savoye et al.
Protein expression and purification, 30(1), 18-25 (2003-06-25)
FtsW and RodA are homologous integral membrane proteins involved in bacterial cell division and cell growth, respectively. Both proteins from Streptococcus pneumoniae were overexpressed in Escherichia coli as N-terminal His-tagged fusions. Their membrane addressing in E. coli was demonstrated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service