Skip to Content
Merck
All Photos(1)

Key Documents

301M-1

Sigma-Aldrich

Cytokeratin, LMW (AE1) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

AE1, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (301M-14)
vial of 0.5 mL concentrate (301M-15)
bottle of 1.0 mL predilute (301M-17)
vial of 1.0 mL concentrate (301M-16)
bottle of 7.0 mL predilute (301M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1κ

control

prostate

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... KRT1(3848)

General description

Anti-cytokeratin, low molecular weight (AE1) labels most acidic keratins; therefore, it is a broadly reactive antibody staining most epithelia and their neoplasms. Members of the acidic and basic cytokeratin subfamilies are found together in pairs; each epithelium contains at least one acidic and one basic keratin so this antibody can show the distribution of keratin containing cells in epithelia. This antibody has shown great sensitivity and broad specificity for keratins under various conditions of fixation and staining. Anti-low molecular weight cytokeratin (AE1) is particularly suited to distinguish poorly differentiated carcinomas from non-epithelial neoplasms. This marker stains both normal and neoplastic cells of epithelial origin.

Quality


IVD

IVD

IVD

RUO

Linkage

Cytokeratin, LMW Positive Control Slides, Product No. 301S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Demetris et al.
The American journal of pathology, 149(2), 439-448 (1996-08-01)
The ductular reaction to acute submassive necrosis was studied in human livers removed at the time of orthotopic liver transplantation. Single, double, and triple immunohistochemical labeling in combination with morphometry was used to analyze the phenotype and proliferative and apoptotic
Diagnostic Immunohistochemistry: Theranostic and Genomic Applications, 166-174 (2002)
R Kitazawa et al.
Virchows Archiv : an international journal of pathology, 435(2), 137-142 (1999-12-22)
A 69-year-old man had a hepatic tumour occupying the left and half of the right lobe, with portal vein thrombus. There were hypercalcaemia and hypophosphataemia with increased nephrogenous cyclic adenosine monophosphate; bone metastases were excluded. Serum parathyroid hormone-related protein (PTHrP)
A R Judkins et al.
American journal of clinical pathology, 110(5), 641-646 (1998-11-05)
Immunohistochemistry occasionally is used to determine the lineage of entirely necrotic tumors. However, the sensitivity and specificity of antibodies on necrotic tissue are unknown. To determine the usefulness of immunohistochemistry with necrotic lesions, a series of 24 known tumors consisting
P E Swanson
American journal of clinical pathology, 95(4 Suppl 1), S2-S7 (1991-04-01)
Historically, antibodies to cytokeratin intermediate filaments have been models of target specificity. In most diagnostic settings, the utility of these antibodies was unquestioned; reactivity for cytokeratin was dogmatically equated with epithelial differentiation. Recently, however, the diagnostic importance of these antibodies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service