Skip to Content
Merck
All Photos(1)

Key Documents

49975

Supelco

Acacetin

analytical standard

Synonym(s):

4′-Methylapigenin, 5,7-Dihydroxy-4′-methoxyflavone, Apigenin 4′-methyl ether, Buddleoflavonol, Linarigenin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
Beilstein:
277879
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤5.0% water

mp

260-265 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C2=CC(=O)c3c(O)cc(O)cc3O2

InChI

1S/C16H12O5/c1-20-11-4-2-9(3-5-11)14-8-13(19)16-12(18)6-10(17)7-15(16)21-14/h2-8,17-18H,1H3

InChI key

DANYIYRPLHHOCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Flavonoid with anti-peroxidative and anti-inflammatory properties. Used in traditional Chinese medicine. It has been recognized as an antiproliferative agent on cancer cell lines by inducing apoptosis and blocking cell cycle progression in the G1 phase.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service