Skip to Content
Merck
All Photos(2)

Key Documents

MABN263

Sigma-Aldrich

Anti-GABA(A) receptor subunit gamma-2 Antibody, clone 10F10-C1-B8

clone 10F10-C1-B8, from mouse

Synonym(s):

Gamma-aminobutyric acid receptor subunit gamma-2, GABA(A) receptor subunit gamma-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10F10-C1-B8, monoclonal

species reactivity

rat

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

GABA(A) receptor subunit gamma-2 is a protein belonging to a subclass of GABA receptors known as GABA (A). GABA (A) receptors are pentameric chloride channels composed of subunits belonging to different homology classes, i.e. α, β, γ, δ, θ, ε, π, and act as receptors for GABA, a crucial inhibitory neurotransmitter localized within the brain. GABA (A) gamma 2 subunit consists of 5 cysteine residues and carries the binding site for benzodiazepine. The subunit functions to promote receptor maintenance and clustering at the synapse, interacts with GABA receptor associated proteins such as GODZ and GABAWRAP, affects GABA (A) receptor conductance and kinetics, and is a contributing component to receptors participating in both phasic and tonic inhibition. Several mutations in the GABA(A) receptor subunit gamma-2 expression have been linked to epileptic syndromes that result in fibrile seizures.

Immunogen

Linear peptide corresponding to rat GABA(A) receptor subunit gamma-2.

Application

Detect GABA(A) receptor subunit gamma-2 using this Anti-GABA(A) receptor subunit gamma-2 Antibody, clone 10F10-C1-B8 validated for use in Western Blotting, ICC.
Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected GABA(A) receptor subunit gamma-2 in rat hippocampal neurons. Image courtesy from the laboratory of Dr. Jaideep Kapur, University of Virginia.

Quality

Evaluated by Western Blot in PC-12 cell lysate.

Western Blot Analysis: A 1:2,000 dilution of this antibody detected GABA(A) receptor subunit gamma-2 in 10 µg of PC-12 cell lysate.

Target description

~54 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service