Skip to Content
Merck
All Photos(1)

Key Documents

AB5340P

Sigma-Aldrich

Anti-Cocaine and Amphetamine Related Transcript Antibody

Chemicon®, from chicken

Synonym(s):

CART/CARP

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€692.00

€692.00


Estimated to ship on18 April 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€692.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€692.00


Estimated to ship on18 April 2025


Request a Bulk Order

biological source

chicken

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgY

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

Recognizes Cocaine and Amphetamine Related Transcript (CART). CART has been found to control satiety modulating the actions of two key regulators of food intake, leptin and NPY. Starvation decreases CART levels in the arcuate nucleus (Kristensen et al. 1998). Obese animals have virtually no CART. CART inhibits both normal and starvation-induced feeding, and completely inhibits NPY-induced feeding. Immunoneutralization of CART by administration of anti-CART resulted into higher food intake suggesting that CART is an endogenous regulator of food intake.

Immunogen

Mouse/rat CART peptide (55-102).

Application

Detect Cocaine & Amphetamine Related Transcript using this Anti-Cocaine & Amphetamine Related Transcript Antibody validated for use in ELISA, WB, IH.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Reward & Addiction
Western blotting: 1-10 μg/mL using ECL.

Immunohistochemistry: It has been reported that the antibody will recognize CART in neurons (cell bodies) from the rat hypothalamic arcuate nucleus.

ELISA: 1:10,000-1:50,000 using 50-100 ng control peptide per well.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS with 0.1% BSA

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Bäckberg et al.
Journal of neuroendocrinology, 16(7), 589-604 (2004-06-25)
gamma-Aminobutyric acid (GABA), the major inhibitory neurotransmitter in the brain, acts via two different type of GABA receptors. GABA(A) receptors are composed of five subunits that belong to eight different classes. Depending on their subunit composition, distinct pharmacological and electrophysiological
M Bäckberg et al.
Journal of neuroendocrinology, 15(1), 1-14 (2003-01-22)
Gamma-aminobutyric acid (GABA) interacts with hypothalamic neuronal pathways regulating feeding behaviour. GABA has been reported to stimulate feeding via both ionotropic GABA(A) and metabotropic GABA(B) receptors. The functional form of the GABA(B) receptor is a heterodimer consisting of GABA(B) receptor-1
Yu Kodani et al.
eNeuro, 9(2) (2022-04-20)
Hypothalamic melanin-concentrating hormone (MCH) neurons are important regulators of multiple physiological processes, such as sleep, feeding, and memory. Despite the increasing interest in their neuronal functions, the molecular mechanism underlying MCH neuron development remains poorly understood. We report that a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service