Skip to Content
Merck
All Photos(1)

Documents

ENAH5802DA23

Enamine

2,2-dimethyl-4-[2-(prop-2-yn-1-yloxy)ethyl]thiomorpholine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
213.34
MDL number:

Iupac Name

2,2-dimethyl-4-(2-(prop-2-yn-1-yloxy)ethyl)thiomorpholine

Inchi Code

1S/C11H19NOS/c1-4-7-13-8-5-12-6-9-14-11(2,3)10-12/h1H,5-10H2,2-3H3

InChI key

NJFCUCFXLWQJMI-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-6063131

MSDS

https://www.enaminestore.com/catalog/EN300-6063131


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service