Skip to Content
Merck
All Photos(1)

Key Documents

ENA424045512

Enamine

3-bromo-1-(2,4-dichlorophenyl)pyrrolidin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
308.99
MDL number:

Iupac Name

3-bromo-1-(2,4-dichlorophenyl)-2-pyrrolidinone

Inchi Code

1S/C10H8BrCl2NO/c11-7-3-4-14(10(7)15)9-2-1-6(12)5-8(9)13/h1-2,5,7H,3-4H2

InChI key

XJBYHOVONSVERO-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-174720

MSDS

https://www.enaminestore.com/catalog/EN300-174720


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service