Skip to Content
Merck
All Photos(1)

Key Documents

ATEH57C9A1D4

AstaTech, Inc.

CIS-6-BENZYLOCTAHYDRO-1H-PYRROLO[3,4-B]PYRIDINE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
216.33
MDL number:

Iupac Name

(4aR,7aR)-6-benzyloctahydro-1H-pyrrolo[3,4-b]pyridine

Inchi Code

1S/C14H20N2/c1-2-5-12(6-3-1)9-16-10-13-7-4-8-15-14(13)11-16/h1-3,5-6,13-15H,4,7-11H2/t13-,14+/m1/s1

InChI key

AFYZAHZKOFBVLE-KGLIPLIRSA-N

Purity

95%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=123403

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=W14366


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service