Skip to Content
Merck
All Photos(1)

Key Documents

APO455837973

Apollo Scientific Ltd

2,2,2-Trifluoroethyl trichloromethanesulphonate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€86.20

Vendor SKU: PC7377-1G

€86.20


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
€86.20

About This Item

CAS Number:
Molecular Weight:
281.47
MDL number:

Vendor SKU: PC7377-1G

€86.20


Ships in 14 days - from Aldrich Partner.

bp

-70

density

1.714

Iupac Name

2,2,2-trifluoroethyl trichloromethanesulfonate

Inchi Code

1S/C3H2Cl3F3O3S/c4-3(5,6)13(10,11)12-1-2(7,8)9/h1H2

InChI key

GOIWZZQXWJVDOG-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/222-trifluoroethyl-trichloromethanesulphonate

Flash Point

84-86/20mm

Boiling point

-70

density

1.714

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/222-trifluoroethyl-trichloromethanesulphonate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service