AMBH97F07673
Ambeed, Inc.
3-Oxo-3-(thiophen-3-yl)propanenitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
3-oxo-3-(3-thienyl)propanenitrile
Inchi Code
1S/C7H5NOS/c8-3-1-7(9)6-2-4-10-5-6/h2,4-5H,1H2
InChI key
IWPZODDPZNLDAM-UHFFFAOYSA-N
storage temp.
Keep in dark place,Sealed in dry,2-8C
Purity
95%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
solid
Product Link
https://www.ambeed.com/products/69879-30-7.html
Signal Word
Warning
Hazard Statements
MSDS
https://www.ambeed.com/products/69879-30-7.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service