Key Documents
Z136255
Tubing sizer
determines I.D. and O.D. of glass tubing, test tubes, etc
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
material
polypropylene
manufacturer/tradename
Bel-Art
measuring range
1.5-16 mm
L × W × H
120 mm × 50 mm × 12 mm
General description
Determines I.D. and O.D. of glass tubing, test tubes, etc. For I.D., select the closest pin size; for O.D., choose closest well size.
Choose from one of the most recent versions:
Certificates of Analysis (COA)