Skip to Content
Merck
All Photos(1)

Documents

W379204

Sigma-Aldrich

2-Pentanethiol

≥97%

Synonym(s):

2-Mercaptopentane, sec-Amyl mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(SH)CH3
CAS Number:
Molecular Weight:
104.21
FEMA Number:
3792
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
12.192

Assay

≥97%

refractive index

n20/D 1.4410 (lit.)

bp

101 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

SMILES string

CCCC(C)S

Features and Benefits

Unpleasant odor, sulfuraceous, gassy

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service