Documents
BAF420310000
SP Bel-Art® "Space Saver" Polycarbonate Vacuum Desiccator
volume 0.20 ft3
Synonym(s):
SP Bel-Art® "Space Saver" Polycarbonate Vacuum Desiccator
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
material
clear polycarbonate top(s)
white polypropylene bottom
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F42031-0000
volume
0.20 ft3
General description
Maximize your space in the laboratory or classroom. In less than 12" of benchtop space, these strong, shatterproof desiccators will provide dry storage and protection for moisture-sensitive products.
  • ‘Flat Dome,’ clear polycarbonate top with knob maximizes interior clearances and provides an average of 13% greater interior volume over standard hemispherical domes
  • Large ribbed knob for easy lifting of top; extra heavy walls
  • All models include a 0.32 cm (⅛") thick perforated plate; disposable or reusable desiccant can be placed on or under this plate
  • A neoprene O-ring assures a tight seal down to 0˚C without the need for grease (ring will not be harmed if grease is used.)
  • White polypropylene bottom can support incandescent crucibles on High Heat Minerit HD Ceramic Desiccator Plates
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP