Skip to Content
Merck
All Photos(1)

Documents

A5802

Sigma-Aldrich

Aminoacetonitrile

≥98%

Synonym(s):

Cyanomethylamine, Glycinonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4N2
CAS Number:
Molecular Weight:
56.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

liquid

storage temp.

−20°C

SMILES string

NCC#N

InChI

1S/C2H4N2/c3-1-2-4/h1,3H2

InChI key

DFNYGALUNNFWKJ-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Bumsup Kwon et al.
Neurobiology of learning and memory, 97(1), 132-139 (2011-11-17)
The intracellular mitogen-activated protein kinase (MAPK) pathway in the brain is necessary for the formation of a variety of memories including conditioned taste aversion (CTA) learning. However, the functional role of MAPK activation in the amygdala during lithium chloride (LiCl)-induced
K E Baker et al.
Veterinary parasitology, 186(3-4), 513-517 (2011-12-16)
Three experiments defined the resistance profile of a population of Haemonchus contortus, which was shown to express multiple resistances to the benzimidazole, levamisole, macrocyclic lactone and salicylanilide anthelmintic classes when given as a registered combination. Study 1 was a faecal
Establishing an efficacious dose rate of monepantel for treating gastrointestinal nematodes in llamas under field conditions.
A M Dadak et al.
The Veterinary record, 172(6), 155-155 (2013-01-08)
Lucie Stuchlíková et al.
Analytical and bioanalytical chemistry, 405(5), 1705-1712 (2012-11-28)
Monepantel (MOP) belongs to a new class of anthelmintic drugs known as aminoacetonitrile derivatives. It was approved for use in veterinary practice in Czech Republic in 2011. So far, biotransformation and transport of MOP in target animals have been studied
P A Stein et al.
Veterinary parasitology, 185(2-4), 339-342 (2011-11-15)
A controlled, blinded study was undertaken in 6-week old, pre-weaned lambs to demonstrate the safety and efficacy against fourth-stage gastrointestinal nematode larvae, of monepantel administered per os at 2.5mg/kg body weight. Worm burdens of 10 monepantel-treated lambs were compared to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service