Skip to Content
Merck
All Photos(1)

Key Documents

772925

Sigma-Aldrich

4,6-O-Isopropylidene-D-glucal

97%

Synonym(s):

1,5-Anhydro-2-deoxy-4,6-O-(1-methylethylidene)-D-arabino-hex-1-enitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O4
CAS Number:
Molecular Weight:
186.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.487

density

1.179 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

O[C@@H]1C=CO[C@@]2([H])[C@@]1([H])OC(C)(C)OC2

InChI

1S/C9H14O4/c1-9(2)12-5-7-8(13-9)6(10)3-4-11-7/h3-4,6-8,10H,5H2,1-2H3/t6-,7-,8+/m1/s1

InChI key

GAQDIYMHBQNXLE-PRJMDXOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service