Skip to Content
Merck
All Photos(1)

Key Documents

738344

Sigma-Aldrich

Naphthalene-1-thiocarboxamide

97%

Synonym(s):

Naphthalene-1-carbothioamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9NS
CAS Number:
Molecular Weight:
187.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

126-130 °C

SMILES string

NC(=S)c1cccc2ccccc12

InChI

1S/C11H9NS/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h1-7H,(H2,12,13)

InChI key

DRCKUACWKCMOCB-UHFFFAOYSA-N

Application

Naphthalene-1-thiocarboxamide is an organosulphur compound, which may be used in dehydrosulfurization of aromatic thioamides to nitriles.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dehydrosulfurization of Aromatic Thioamides to Nitriles Using Indium (III) Triflate.
Mineno T, et al.
International Journal of Organic Chemistry, 4(03), 169-169 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service