Skip to Content
Merck
All Photos(1)

Documents

727598

Sigma-Aldrich

2-Aminobenzoxazole

≥97.0%

Synonym(s):

2-Benzoxazolamine, Benzooxazol-2-ylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N2O
CAS Number:
Molecular Weight:
134.14
Beilstein:
115981
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (GC)
≥97.0%
97.0-103.0% (NT)

mp

128-133 °C

SMILES string

Nc1nc2ccccc2o1

InChI

1S/C7H6N2O/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H,(H2,8,9)

InChI key

JPBLHOJFMBOCAF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service