Skip to Content
Merck
All Photos(1)

Key Documents

656267

Sigma-Aldrich

Poly(9,9-di-(2-ethylhexyl)-9H-fluorene-2,7-vinylene)

Synonym(s):

PFV

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C31H42)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

transition temp

Tg (DSC) 173 °C (Lit.)(lit.)

fluorescence

λex 281 nm; λem 454 nm in chloroform
λem 507 nm (Film Photoluminescence)(lit.)

Application

Poly(9,9-di-(2-ethylhexyl)-9H-fluorene-2,7-vinylene) is a fluorescent conjugating polymer which can be combined with nanoparticles to form a nanocomposite for optoelectronics applications. It can also be used in the formation of a cathode material for organic light emitting diodes (OLEDs).[1][2]

Features and Benefits

A green-blue polymer for OLEDs with good turn on voltage (2.6V) and maximum brightness and luminecence effieciency.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optical Studies of Poly (9, 9-di-(2-ethylhexyl)-9H-fluorene-2, 7-vinylene) and its Nanocomposites
Layek S, et al.
Journal of Applied Spectroscopy, 82(5), 868-874 (2015)
Influence of poly (3, 4-ethylenedioxythiophene)-poly (styrenesulfonate) in polyfluorene-based light-emitting diodes: Evidence of charge trapping at the organic interface
Kanaan H, et al.
Organic Electronics, 11(6), 1047-1052 (2010)
Jin, S.-H.; Kang, S.-Y.; Kim, M.-Y.; Chang, Y.U.
Macromolecules, 36, 3841-3841 (2003)

Questions

1–2 of 2 Questions  
  1. Is this polymer R, S, or Racemate?

    1 answer
    1. The polymer is racemic, and whether it is R or S would depend on the monomer, and that information in unavailable.

      Helpful?

  2. Is this polymer R, S or Racemate?

    1 answer
    1. This is a racemic polymer.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service