Skip to Content
Merck
All Photos(2)

Key Documents

378216

Sigma-Aldrich

1-Fluoro-2,4,6-trimethylpyridinium triflate

technical grade, 85%

Synonym(s):

1-Fluoro-sym-collidinium triflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11F4NO3S
CAS Number:
Molecular Weight:
289.25
Beilstein:
4777118
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

85%

mp

162-165 °C (lit.)

SMILES string

[O-]S(=O)(=O)C(F)(F)F.Cc1cc(C)[n+](F)c(C)c1

InChI

1S/C8H11FN.CHF3O3S/c1-6-4-7(2)10(9)8(3)5-6;2-1(3,4)8(5,6)7/h4-5H,1-3H3;(H,5,6,7)/q+1;/p-1

InChI key

PRIGFEJKMMRJSF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Pd(II)-catalyzed para-selective C-H arylation
  • Fluorination reactions
  • Oxidant in oxidatively-induced aryl-CF3 bond-formation
  • Pd(II)-catalyzed C-H olefination reactions

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service