Skip to Content
Merck
All Photos(1)

Key Documents

246042

Sigma-Aldrich

3-Butyn-2-ol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CCH(OH)CH3
CAS Number:
Molecular Weight:
70.09
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

98%

refractive index

n20/D 1.426 (lit.)

bp

66-67 °C/150 mmHg (lit.)

density

0.894 g/mL at 25 °C (lit.)

SMILES string

CC(O)C#C

InChI

1S/C4H6O/c1-3-4(2)5/h1,4-5H,2H3

InChI key

GKPOMITUDGXOSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service