Skip to Content
Merck
All Photos(1)

Key Documents

237574

Sigma-Aldrich

1,4-Dibromo-2,3-butanediol

95%, mixture of R,R and S,S isomers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH(OH)CH(OH)CH2Br
CAS Number:
Molecular Weight:
247.91
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

82-84 °C (lit.)

SMILES string

BrCC(O)C(O)CBr

InChI

1S/C4H8Br2O2/c5-1-3(7)4(8)2-6/h3-4,7-8H,1-2H2/t3-,4-/m1/s1

InChI key

XOWDQAHYPSENAC-QWWZWVQMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Dibromo-2,3-butanediol reacts with aqueous alkaline sodium arsenite to yield mixture of DL-2,3,4-trihydroxybutylarsonic acid and DL-2,3-dihydroxybutane-1,4-bis(arsonic acid).

Application

1,4-Dibromo-2,3-butanediol has been used in the preparation of diquaternary gemini surfactants.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerasimos M Tsivgoulis et al.
Chemistry and physics of lipids, 148(2), 97-104 (2007-06-09)
The reaction of DL-1,3-butadiene diepoxide and of DL-1,4-dibromo-2,3-butanediol with aqueous alkaline sodium arsenite, "Na(3)AsO(3)", gave mixtures of the title arsonic acids which can be separated by anion exchange resin. Characterization of by-products leads to a better understanding of these reactions.
Riikka Mäkelä et al.
The Journal of clinical endocrinology and metabolism, 88(8), 3823-3828 (2003-08-14)
Myeloperoxidase (MPO) is an oxidative enzyme present in phagocytes and atherosclerotic lesions. The MPO gene has a promoter polymorphism -463G/A, which leads to high (GG) and low expression (AG, AA) genotypes. We investigated the effect of long-term hormone replacement therapy
A M Kessling et al.
American journal of human genetics, 42(3), 458-467 (1988-03-01)
We have investigated the frequencies of RFLPs of the apolipoprotein (apo) AII gene and of the apo AI-CIII-AIV gene cluster in 109 men, selected from a random sample of 1,910 men aged 45-59 years, to cover a wide range of
Surface activity and premicellar aggregation of some novel diquaternary gemini surfactants.
Rosen MJ and Liu L.
Journal of the American Oil Chemists' Society, 73(7), 885-890 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service