Skip to Content
Merck
All Photos(1)

Documents

18853

Sigma-Aldrich

1,2-Butadiene

≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C=CHCH3
CAS Number:
Molecular Weight:
54.09
Beilstein:
1730808
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:

Assay

≥99.0% (GC)

technique(s)

NMR: suitable (13C-NMR)

bp

10.8 °C (lit.)

SMILES string

CC=C=C

InChI

1S/C4H6/c1-3-4-2/h4H,1H2,2H3

InChI key

QNRMTGGDHLBXQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

pressure tin

Other Notes

Sales restrictions may apply

control valve

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ralf I Kaiser et al.
The journal of physical chemistry. A, 118(32), 6181-6190 (2014-08-02)
The reactions of the 4-tolyl radical (C6H4CH3) and of the D7-4-tolyl radical (C6D4CD3) with 1,2-butadiene (C4H6) have been probed in crossed molecular beams under single collision conditions at a collision energy of about 54 kJ mol(-1) and studied theoretically using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service