Skip to Content
Merck
All Photos(1)

Documents

137022

Sigma-Aldrich

1,2-Diphenylethylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(C6H5)NH2
CAS Number:
Molecular Weight:
197.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

EMPROVE® Bio

Assay

97%

form

liquid

refractive index

n20/D 1.58 (lit.)

bp

310-311 °C/750 mmHg (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

NC(Cc1ccccc1)c2ccccc2

InChI

1S/C14H15N/c15-14(13-9-5-2-6-10-13)11-12-7-3-1-4-8-12/h1-10,14H,11,15H2

InChI key

DTGGNTMERRTPLR-UHFFFAOYSA-N

General description

(S)- and (R)-enantiomers of 1,2-diphenylethylamine are the precursors for synthesis of (S)- and (R)-1-(1,2-diphenylethyl)piperidine.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael L Berger et al.
Bioorganic & medicinal chemistry, 17(9), 3456-3462 (2009-04-07)
We resolved 1,2-diphenylethylamine (DPEA) into its (S)- and (R)-enantiomer and used them as precursors for synthesis of (S)- and (R)-1-(1,2-diphenylethyl)piperidine, flexible homeomorphs of the NMDA channel blocker MK-801. We also describe the synthesis of the dicyclohexyl analogues of DPEA. These
Carina S D Wink et al.
Analytical and bioanalytical chemistry, 407(6), 1545-1557 (2015-01-13)
Lefetamine (N,N-dimethyl-1,2-diphenylethylamine, L-SPA) was marketed as an opioid analgesic in Japan and Italy. After being widely abused, it became a controlled substance. It seems to be a pharmaceutical lead for designer drugs because N-ethyl-1,2-diphenylethylamine (NEDPA) and N-iso-propyl-1,2-diphenylethylamine (NPDPA) were confiscated
Daniel A Spudeit et al.
Journal of chromatography. A, 1363, 89-95 (2014-08-30)
This work reports a comparison of HPLC separations of enantiomers with chiral stationary phases (CSPs) prepared by chemically bonding cyclofructan-6, functionalized with isopropyl carbamate groups on fully and superficially porous particles (SPPs). The chromatographic performance of the superficially porous CSP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service