Skip to Content
Merck
All Photos(1)

Documents

112240

Sigma-Aldrich

Cyclododecanemethanol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H26O
CAS Number:
Molecular Weight:
198.34
MDL number:

Assay

95%

SMILES string

OCC1CCCCCCCCCCC1

InChI

1S/C13H26O/c14-12-13-10-8-6-4-2-1-3-5-7-9-11-13/h13-14H,1-12H2

InChI key

JQYWAZNRRQRPNN-UHFFFAOYSA-N


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service