Skip to Content
Merck
All Photos(1)

Key Documents

M3553

Sigma-Aldrich

EX-CELL® MDBK-GM Medium

with HEPES, with sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

MDBK-GM Serum-free Medium

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352200

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

storage temp.

2-8°C

Application

Formulated to support the growth of Madin-Darby Bovine Kidney (MDBK) cells for the production of viruses. Designed to be used with M 0682.

Components

Complete medium containing 150 μg/ml animal-derived protein, inorganic salts, HEPES and sodium bicarbonate buffers, essential and non-essential amino acids, vitamins, recombinant human insulin and growth factors, fetuin, transferrin, other organic compounds, and trace elements. Without L-glutamine

Reconstitution

Aseptically add 16.5 ml of 200 mM L-glutamine solution per liter of medium prior to use.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service