Merck
3D Printing
Stereolithography 3D printer showing 3D object emerging from vat of resin
3D printing, also called additive manufacturing, is a process for making a three-dimensional object from a digital model by successively fabricating material layer-by-layer in an additive manner. First, a virtual 3D representation of the object is rendered using computer-aided design (CAD). Then, the model is ‘sliced’ into a series of horizontal layers to convert the design into an STL (standard tessellation language) file that is readable for the 3D printer. This data is then transferred to the printer, and the printer settings are defined. The final object is produced one layer at a time, with each layer bonding and building on the previous one.  
Due to the ability to produce very complex shapes and structures with high precision and repeatability from a diverse range of materials, 3D printing is used for a broad range of applications in aerospace, automotive, construction, fashion, food, jewelry, manufacturing, and medical sectors. There are numerous techniques to 3D print objects with different mechanical, thermal, and chemical properties from materials in molten, liquid, or powder states.
For more in-depth information, including minimizing your carbon footprint with smart manufacturing, please explore our 3D to 4D Printing for Added Sustainability Brochure.
Among the popular 3D printing technologies are:​
Related Technical Articles
  • Partnering additive manufacturing (3D printing) with functional nanomaterial-based inks has the potential to push the properties and performance of advanced materials beyond previous capabilities. This is particularly true in energy and environmental applications.
  • A nanocomposite is typically defined as a mixture between a host material (e.g., polymer matrix) and nanofillers with at least one dimension of less than 100 nm.
  • Three-dimensional (3D) printing technology, also called additive manufacturing (AM), has recently come into the spotlight because of its potential high-impact implementation in applications ranging from personal tools to aerospace equipment.
  • The emerging field of printed electronics requires a suite of functional materials for applications including flexible and large-area displays, radio frequency identification tags, portable energy harvesting and storage, biomedical and environmental sensor arrays,5,6 and logic circuits.
  • Inkjet printing is one of the key enabling technologies of printed electronics. Inkjet printing technology classification, aspects of materials (inks, substrates) and respective pre-and post-processing steps are discussed.
  • See All (21)
Find More Articles
Vat Polymerization 
Vat polymerization uses photopolymerization to cure and solidify liquid polymer resin. Stereolithography (SL) was the first type of 3D printing developed and commercialized using this method. An SL printer uses mirrors positioned at the X-Y axes to direct a laser beam across a vat of resin to create a cross-section of the object. Digital light processing (DLP) uses a projector with an LCD screen or UV light source to flash light, creating each layer. This type of resin-based 3D printing is fast because an entire layer is exposed all at once. Masked stereolithography (MLA) uses an array of LEDs to shine UV light through a liquid crystal display (LCD) photomask.
Fused Deposition Modeling(FDM)
Fused deposition modeling (FDM), also called fused filament fabrication (FFF) or material extrusion, is the most common and inexpensive 3D printing technology. A spool of thermoplastic filament (e.g., PLA, ABS) is heated to its melting point and extruded through a nozzle onto a platform, where the molten material cools and solidifies. This technology is used in injection molding and modern plastic manufacturing for ready-to-use products.
Powder Bed Fusion 
Powder bed fusion selectively cures polymer or metal powders with a thermal energy source to create a solid plastic or metal object. First, the powder is heated to a temperature just below its melting point. Then, a roller distributes a very thin layer of powder over the surface of the building bed before a laser passes over the layer to fuse it. Once a layer is completed, the powder bed sinks incrementally to form the next layer. Selective laser sintering (SLS) successively sinters polymer powder with a laser. Selective laser melting (SLM) involves fully melting metal powder instead of sintering it. Other forms of metal powder bed fusion include direct metal laser sintering (DMLS) and electron beam melting (EBM). 
Jetting
Material jetting uses inkjet printer technology to add tiny droplets of photopolymers or wax onto a build plate. An ultraviolet (UV) light simultaneously cures the layers as they are printed. Material jetting (MJ) deposits material in a rapid, line-wise fashion, rather than point-wise. Therefore, multiple objects can be fabricated in a single line. Moreover, this method allows different materials to be printed in the same object. Drop-on-demand (DOD) 3D printing technology uses a set of two inkjets to deposit both the final object material and dissolvable support material simultaneously.
Highlights
Graphene and New Monoatomic Materials
Graphene & New Monoatomic Materials
Join Professor Palermo to learn more about using 2D nanosheets in a 3D world.
3D-Printable Biomaterial Ink Platforms Webinar
Learn more about hydrogel bioprinting and 3D-painting in this webinar.