Merck
  • Inactivation of the Na,K-ATPase by modification of Lys-501 with 4-acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid (SITS).
Inactivation of the Na,K-ATPase by modification of Lys-501 with 4-acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid (SITS).
FEBS letters (1992-12-07)
C H Pedemonte, T L Kirley, M J Treuheit, J H Kaplan
ZUSAMMENFASSUNG
The sodium pump or Na,K-ATPase, maintains the Na+ and K+ gradients across eukaryotic cell membranes at the expense of ATP. Incubation of purified canine renal Na,K-ATPase with 4-acetamido-4'-isothiocyanatostilbene-2,2'-disulfonic acid (SITS) inhibited the ATPase activity. Both the labeling of the protein and the loss of ATPase activity were prevented by co-incubation with ADP (acting as an ATP analog) or KCl. Only the alpha-subunit was labeled by SITS. The alpha-subunit from the inhibited enzyme was extensively digested with trypsin, and SITS-labeled peptides were purified by reverse-phase HPLC and sequenced. The amino acid sequence determined, His-Leu-Leu-Val-Met-X-Gly-Ala-Pro-Glu, indicated that SITS modifies Lys-501 (X) on the alpha-subunit of Na,K-ATPase.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Acetamido-4′-Isothiocyanato-2,2′-Stilbenedisulfonsäure Dinatriumsalz, ≥80%