Merck
Specific inhibitors of poly(ADP-ribose) synthetase and mono(ADP-ribosyl)transferase.
The Journal of biological chemistry (1992-01-25)
M Banasik, H Komura, M Shimoyama, K Ueda
ZUSAMMENFASSUNG
Two classes of enzymes, poly(ADP-ribose) synthetase and mono(ADP-ribosyl)transferases, catalyze covalent attachment of multiple or single residues, respectively, of the ADP-ribose moiety of NAD+ to various proteins. In order to find good inhibitors of poly(ADP-ribose) synthetase free of side actions and applicable to in vivo studies, we made a large scale survey using an in vitro assay system, and found many potent inhibitors. The four strongest were 4-amino-1,8-naphthalimide, 6(5H)- and 2-nitro-6(5H)-phenanthridinones, and 1,5-dihydroxyisoquinoline. Their 50% inhibitory concentrations, 0.18-0.39 microM, were about two orders of magnitude lower than that of 3-aminobenzamide that is currently most popularly used. A common structural feature among all potent inhibitors, including 1-hydroxyisoquinoline, chlorthenoxazin, 3-hydroxybenzamide, and 4-hydroxyquinazoline, in addition to the four mentioned above, was the presence of a carbonyl group built in a polyaromatic heterocyclic skeleton or a carbamoyl group attached to an aromatic ring. Most of the inhibitors exhibited mixed-type inhibition with respect to NAD+. Comparative studies of the effects on poly(ADP-ribose) synthetase and mono(ADP-ribosyl)transferase from hen heterophils revealed high specificity of most of the potent inhibitors for poly(ADP-ribose) synthetase. On the other hand, unsaturated long-chain fatty acids inhibited both enzymes, and saturated long-chain fatty acids and vitamin K1 acted selectively on mono(ADP-ribosyl)transferase. The finding of many inhibitors of ADP-ribosyltransferases, especially poly(ADP-ribose) synthetase, supports the view that ADP-ribosylation of proteins may be regulated by a variety of metabolites or structural constituents in the cell.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Amino-1,8-Naphthalimid