Merck
Looking for Missing Proteins in the Proteome of Human Spermatozoa: An Update.
Journal of proteome research (2016-11-05)
Yves Vandenbrouck, Lydie Lane, Christine Carapito, Paula Duek, Karine Rondel, Christophe Bruley, Charlotte Macron, Anne Gonzalez de Peredo, Yohann Couté, Karima Chaoui, Emmanuelle Com, Alain Gateau, Anne-Marie Hesse, Marlene Marcellin, Loren Méar, Emmanuelle Mouton-Barbosa, Thibault Robin, Odile Burlet-Schiltz, Sarah Cianferani, Myriam Ferro, Thomas Fréour, Cecilia Lindskog, Jérôme Garin, Charles Pineau
ZUSAMMENFASSUNG
The Chromosome-Centric Human Proteome Project (C-HPP) aims to identify "missing" proteins in the neXtProt knowledgebase. We present an in-depth proteomics analysis of the human sperm proteome to identify testis-enriched missing proteins. Using protein extraction procedures and LC-MS/MS analysis, we detected 235 proteins (PE2-PE4) for which no previous evidence of protein expression was annotated. Through LC-MS/MS and LC-PRM analysis, data mining, and immunohistochemistry, we confirmed the expression of 206 missing proteins (PE2-PE4) in line with current HPP guidelines (version 2.0). Parallel reaction monitoring acquisition and sythetic heavy labeled peptides targeted 36 ≪one-hit wonder≫ candidates selected based on prior peptide spectrum match assessment. 24 were validated with additional predicted and specifically targeted peptides. Evidence was found for 16 more missing proteins using immunohistochemistry on human testis sections. The expression pattern for some of these proteins was specific to the testis, and they could possibly be valuable markers with fertility assessment applications. Strong evidence was also found of four "uncertain" proteins (PE5); their status should be re-examined. We show how using a range of sample preparation techniques combined with MS-based analysis, expert knowledge, and complementary antibody-based techniques can produce data of interest to the community. All MS/MS data are available via ProteomeXchange under identifier PXD003947. In addition to contributing to the C-HPP, we hope these data will stimulate continued exploration of the sperm proteome.