Merck
Filamin-A is required to mediate SST2 effects in pancreatic neuroendocrine tumours.
Endocrine-related cancer (2016-01-07)
Eleonora Vitali, Valeria Cambiaghi, Alessandro Zerbi, Carlo Carnaghi, Piergiuseppe Colombo, Erika Peverelli, Anna Spada, Giovanna Mantovani, Andrea G Lania
ZUSAMMENFASSUNG
Somatostatin receptor type 2 (SST2) is the main pharmacological target of somatostatin (SS) analogues widely used in patients with pancreatic neuroendocrine tumours (P-NETs), this treatment being ineffective in a subset of patients. Since it has been demonstrated that Filamin A (FLNA) is involved in mediating GPCR expression, membrane anchoring and signalling, we investigated the role of this cytoskeleton protein in SST2 expression and signalling, angiogenesis, cell adhesion and cell migration in human P-NETs and in QGP1 cell line. We demonstrated that FLNA silencing was not able to affect SST2 expression in P-NET cells in basal conditions. Conversely, a significant reduction in SST2 expression (-43 ± 21%, P < 0.05 vs untreated cells) was observed in FLNA silenced QGP1 cells after long term SST2 activation with BIM23120. Moreover, the inhibitory effect of BIM23120 on cyclin D1 expression (-46 ± 18%, P < 0.05 vs untreated cells), P-ERK1/2 levels (-42 ± 14%; P < 0.05 vs untreated cells), cAMP accumulation (-24 ± 3%, P < 0.05 vs untreated cells), VEGF expression (-31 ± 5%, P < 0.01 vs untreated cells) and in vitro release (-40 ± 24%, P < 0.05 vs untreated cells) was completely lost after FLNA silencing. Interestingly, BIM23120 promoted cell adhesion (+86 ± 45%, P < 0.05 vs untreated cells) and inhibited cell migration (-24 ± 2%, P < 0.00001 vs untreated cells) in P-NETs cells and these effects were abolished in FLNA silenced cells. In conclusion, we demonstrated that FLNA plays a crucial role in SST2 expression and signalling, angiogenesis, cell adhesion and cell migration in P-NETs and in QGP1 cell line, suggesting a possible role of FLNA in determining the different responsiveness to SS analogues observed in P-NET patients.