Merck
  • West syndrome, microcephaly, grey matter heterotopia and hypoplasia of corpus callosum due to a novel ARFGEF2 mutation.
West syndrome, microcephaly, grey matter heterotopia and hypoplasia of corpus callosum due to a novel ARFGEF2 mutation.
Journal of medical genetics (2013-07-03)
Ehud Banne, Osama Atawneh, Marco Henneke, Knut Brockmann, Jutta Gärtner, Orly Elpeleg, Simon Edvardson
ZUSAMMENFASSUNG
West syndrome (WS) is an epileptic encephalopathy of childhood, defined by the presence of clustered spasms usually occurring before the age of 1 year, hypsarrhythmia on EEG that is notoriously difficult to define, and developmental arrest or regression. The incidence of WS is 1:3200 live births with an aetiology-dependent prognosis. Up to 80% of children with symptomatic WS suffer from mental retardation, and approximately 50% develop Lennox-Gastaut syndrome. Using homozygosity mapping followed by exome sequencing, we identified a ADP-ribosylation factor (ARF) guanine nucleotide-exchange factor two (brefeldin A-inhibited) (ARFGEF2) mutation in five related infants with WS. ARFGEF2 is involved in the activation of ARFs by accelerating replacement of bound guanosine diphosphate (GDP) with Guanosine triphosphate (GTP), and is involved in Golgi transport. A mutation in ARFGEF2 has been previously described only once, causing microcephaly and periventricular heterotopia. Here, we describe a novel ARFGEF2 mutation in five related patients presenting with WS, microcephaly, periventricular heterotopia and thin corpus callosum.