- Electrophilic Aromatic Bromination Using Bromodimethylsulfonium Bromide Generated in Situ.
Electrophilic Aromatic Bromination Using Bromodimethylsulfonium Bromide Generated in Situ.
The Journal of organic chemistry (1997-06-27)
George Majetich, Rodgers Hicks, Steven Reister
PMID11671753
ZUSAMMENFASSUNG
It has been shown that bromodimethylsulfonium bromide, generated in situ by treating dimethyl sulfoxide with aqueous hydrobromic acid, is a milder and more selective reagent for electrophilic aromatic bromination than elemental bromine.