Merck
  • Application of Optimized Vortex-Assisted Surfactant-Enhanced DLLME for Preconcentration of Thymol and Carvacrol, and Their Determination by HPLC-UV: Response Surface Methodology.
Application of Optimized Vortex-Assisted Surfactant-Enhanced DLLME for Preconcentration of Thymol and Carvacrol, and Their Determination by HPLC-UV: Response Surface Methodology.
Journal of chromatographic science (2015-02-01)
Mehrorang Ghaedi, Mostafa Roosta, Saeid Khodadoust, Ali Daneshfar
ZUSAMMENFASSUNG
A novel vortex-assisted surfactant-enhanced dispersive liquid-liquid microextraction combined with high-performance liquid chromatography (VASEDLLME-HPLC) was developed for the determination of thymol and carvacrol (phenolic compound). In this method, the extraction solvent (CHCl3) was dispersed into the aqueous samples via a vortex agitator and addition of the surfactant (Triton X-100). The preliminary experiments were undertaken to select the best extraction solvent and surfactant. The influences of effective variables were investigated using a Plackett-Burman 2(7-4) screening design and then, the significant variables were optimized by using a central composite design combined with desirability function. Working under optimum conditions specified as: 140 µL CHCl3, 0.08% (w/v, Triton X-100), 3 min extraction time, 6 min centrifugation at 4,500 rpm, pH 7, 0.0% (w/v) NaCl permit achievement of high and reasonable linear range over 0.005-4.0 mg L(-1) with R(2) = 0.9998 (n = 10). The separation of thymol and carvacrol was achieved in <14 min using a C18 column and an isocratic binary mobile phase acetonitrile-water (55:45, v/v) with a flow rate of 1.0 mL min(-1). The VASEDLLME is applied for successful determination of carvacrol and thymol in different thyme and pharmaceutical samples with relative standard deviation <4.7% (n = 5).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natriumphosphat, ACS reagent, ≥99.0%
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natronlauge, 50% in H2O
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥98%
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natronlauge, BioUltra, for molecular biology, 10 M in H2O
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natriumphosphat, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, for molecular biology, ≥99%
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, for molecular biology, 10% in H2O
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natriumphosphat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natriumphosphat, BioReagent, for molecular biology, anhydrous, ≥98%
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, for molecular biology, 20% in H2O
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Thymol, ≥98.5%
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge
Sigma-Aldrich
Natronlauge, 5.0 M
SKU
Packungsgröße
Verfügbarkeit
Preis
Menge