Merck
  • Formation of unusual glutamate conjugates of 1-[3-(aminomethyl)phenyl]-N-[3-fluoro-2'-(methylsulfonyl)-[1,1'-biphenyl]-4-yl]-3-(trifluoromethyl)-1H-pyrazole-5-carboxamide (DPC 423) and its analogs: the role of gamma-glutamyltranspeptidase in the biotransformation of benzylamines.
Formation of unusual glutamate conjugates of 1-[3-(aminomethyl)phenyl]-N-[3-fluoro-2'-(methylsulfonyl)-[1,1'-biphenyl]-4-yl]-3-(trifluoromethyl)-1H-pyrazole-5-carboxamide (DPC 423) and its analogs: the role of gamma-glutamyltranspeptidase in the biotransformation of benzylamines.
Drug metabolism and disposition: the biological fate of chemicals (2001-09-19)
A Mutlib, J Shockcor, S Y Chen, R Espina, J Lin, N Graciani, S Prakash, L S Gan
ZUSAMMENFASSUNG
The role of gamma-glutamyltranspeptidase (GGT) in transferring glutamate from endogenous glutathione (GSH) to the benzylamine moiety of a compound, such as 1-[3-(aminomethyl)phenyl]-N-[3-fluoro-2'-(methylsulfonyl)-[1,1'-biphenyl]-4-yl]-3-(trifluoromethyl)-1H-pyrazole-5-carboxamide (DPC 423), is described. Studies were performed with structurally related analogs of DPC 423 to demonstrate that this type of reaction was common to compounds possessing a benzylamine group. Synthesizing appropriate standards and confirming by liquid chromatography (LC)/mass spectroscopy and LC/NMR made unambiguous assignments of the structures of glutamate conjugates of DPC 423. The use of stable isotope-labeled GSH for metabolism studies has not been described before. In the present study, we report the novel use of deuterated GSH in conjunction with mass spectral analysis to demonstrate the glutamate transfer to the benzylamines in the presence of GGT. To further demonstrate that the alpha protons on the benzylamines and glutamate (as part of glutathione) were unaffected during the transpeptidation, these protons were replaced with deuterium. Acivicin (AT-125), a potent and selective inhibitor of GGT, was used to abolish the formation of the glutamate conjugates of DPC 423 in vitro and in vivo. This provided further evidence of the role of GGT in forming the glutamate conjugates of benzylamines. This study demonstrated conclusively that GGT was responsible for mediating the transfer of glutamic acid from GSH to the benzylamine moiety of a series of structurally related compounds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4,4,4-Trifluor-1-(2-furyl)-1,3-butandion, 99%