Merck
  • Bispecific Her2 × cotinine antibody in combination with cotinine-(histidine)2-iodine for the pre-targeting of Her2-positive breast cancer xenografts.
Bispecific Her2 × cotinine antibody in combination with cotinine-(histidine)2-iodine for the pre-targeting of Her2-positive breast cancer xenografts.
Journal of cancer research and clinical oncology (2013-12-03)
Soomin Yoon, Yun-Hee Kim, Se Hun Kang, Seok-Ki Kim, Hwa Kyoung Lee, Hyori Kim, Junho Chung, In-Hoo Kim
ZUSAMMENFASSUNG
Cotinine has optimal characteristics as a hapten for pre-targeted radioimmunotherapy (PRIT). This study was performed to evaluate the applicability of cotinine/anti-cotinine antibody to PRIT. We developed and prepared a tandem, single-chain, variable fragment Fc fusion protein [tandem single-chain variable fragment (scFv) Fc fusion protein] that is reactive to both human epidermal growth factor receptor 2 (Her2) and cotinine. Its simultaneous reactivity to Her2 and cotinine was tested in an enzyme-linked immunosorbent assay (ELISA) and two radioimmunoassays (RIA) employing Her2-coated RIA tubes and a Her2-overexpressing cell line. For in vivo imaging, mice bearing Her2-positive tumors were injected with a mixture of tandem scFv Fc fusion and (125)I-cotinine-conjugated histidine dipeptide ((125)I-cotinine peptide). After a delay, (125)I-cotinine peptide was injected again. ELISA and RIA results showed that tandem scFv Fc fusion protein successfully bound to both Her2 and cotinine. In single-photon emission computed tomography (SPECT), the complex of tandem scFv Fc fusion protein and (125)I-cotinine peptide was localized to Her2-positive tumor xenografts in mice 4 h after the first injection. Enhanced radioactivity at the site of the Her2-positive tumor lesion was monitored 1 h after the second injection. With these findings, we conclude that the tandem scFv Fc fusion protein and cotinine hapten system have the potential to be applied in PRIT.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
trans-4-Cotinincarbonsäure, 97%