Merck
Extending levodopa action: COMT inhibition.
Neurology (1998-06-20)
P Martínez-Martín, C F O'Brien
ZUSAMMENFASSUNG
Degradation of levodopa in the periphery is known to be associated with motor fluctuations and dyskinesia in Parkinson's disease (PD) patients. The enzyme catechol-O-methyltransferase (COMT) is responsible for much of this degradation. Therefore, inhibiting COMT activity is one method of extending the action of levodopa. The new nitrocatechol-type COMT inhibitors entacapone, nitecapone, and tolcapone inhibit COMT in the periphery; tolcapone also inhibits COMT activity centrally. COMT inhibitors increase patients' duration of response to levodopa and reduce response fluctuations. Administration may prolong levodopa-induced dyskinesia, but peak-dose dyskinesia does not appear to increase. To reduce dyskinesia, the total daily dose of levodopa can be reduced.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nitecapone, ≥98% (HPLC)