Merck
  • Synthesis, structure and reactivity of [o-(2,6-diisopropylphenyliminomethinyl)phenyl]selenenyl selenocyanate (RSeSeCN) and related derivatives.
Synthesis, structure and reactivity of [o-(2,6-diisopropylphenyliminomethinyl)phenyl]selenenyl selenocyanate (RSeSeCN) and related derivatives.
Dalton transactions (Cambridge, England : 2003) (2014-03-29)
Prakul Rakesh, Harkesh B Singh, Jerry P Jasinski, James A Golen
ZUSAMMENFASSUNG
The synthesis and the first X-ray structural characterization of a selenenyl selenocyanate, [o-(2,6-diisopropylphenyliminomethinyl)phenyl]selenenyl selenocyanate (), with a stable Se-Se bond are described. The isolation of stable , both in the solid state and in solution, is facilitated by strong intramolecular SeN interaction. The compound , an example of unsymmetrical diselenide, did not exhibit any glutathione peroxidase-like activity. The reaction of with thiophenol afforded (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thiophenol, ≥99%