Merck
  • Synthesis and biological evaluation of trisubstituted imidazole derivatives as inhibitors of p38alpha mitogen-activated protein kinase.
Synthesis and biological evaluation of trisubstituted imidazole derivatives as inhibitors of p38alpha mitogen-activated protein kinase.
Bioorganic & medicinal chemistry letters (2008-06-24)
Dae-Kee Kim, Jin-Hwi Lim, Jung A Lee, Purushottam M Dewang
ZUSAMMENFASSUNG
A series of trisubstituted imidazole derivatives containing a 4-fluorophenyl group, a pyrimidine ring, and a CN- or CONH(2)-substituted benzyl moiety have been synthesized and evaluated for p38alpha MAP kinase inhibitory activity. Among them, compounds 22c, 27b, and 28b inhibited p38alpha MAP kinase with IC(50) values 27.6, 28, and 31 nM, respectively.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methyl-4-fluorbenzoat, 98%