Merck
Freeze-drying above room temperature.
Journal of pharmaceutical sciences (1999-05-07)
M S Tesconi, K Sepassi, S H Yalkowsky
ZUSAMMENFASSUNG
This study investigates the use of solid, organic compounds to lyophilize drugs without conventional freeze-drying equipment. The aim of the investigation is to find a pharmaceutically acceptable solvent or solvent combination that is appropriate for freeze-drying on the basis of its ability to (1) solubilize hydrophobic drugs, (2) provide a stable environment for water-sensitive compounds, (3) be rapidly and completely removed from the product under vacuum, and (4) produce cakes that are readily reconstituted. A eutectic formed from 1,1,1-trichloro-2-methyl-2-propanol (chlorobutanol) hemihydrate and dimethyl sulfone (DMSO2) is determined to be a suitable medium.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,1,1-Trichlor-2-methyl-2-propanol Hemihydrat, 98%