Merck
Synthesis and DP-IV inhibition of cyano-pyrazoline derivatives as potent anti-diabetic agents.
Bioorganic & medicinal chemistry letters (2004-09-11)
Jin Hee Ahn, Hye-Min Kim, Sun Ho Jung, Seung Kyu Kang, Kwang Rok Kim, Sang Dal Rhee, Sung-Don Yang, Hyae Gyeong Cheon, Sung Soo Kim
ZUSAMMENFASSUNG
A new series of cyano-pyrazoline derivatives with secondary amine at P-2 site was synthesized through achiral and chiral synthetic methods and evaluated for their ability to inhibit dipeptidyl peptidase IV (DP-IV). Compound 5i revealed good in vivo efficacy (ED50: 4.1 mg/kg; in vivo DP-IV inhibition). Also chiral derivative (11b) having (S)-configuration of compound 5i was found to be more potent.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloracetylchlorid, 98%