- In vitro evaluation of inflammatory cell response after CF4 plasma surface modification of poly(methyl methacrylate) intraocular lenses.
In vitro evaluation of inflammatory cell response after CF4 plasma surface modification of poly(methyl methacrylate) intraocular lenses.
Journal of cataract and refractive surgery (1993-05-01)
R Eloy, D Parrat, T M Duc, G Legeay, A Bechetoille
PMID8388951
ZUSAMMENFASSUNG
The inflammatory cell response of tetrafluorocarbon (CF4) plasma surface modification of poly(methyl methacrylate) intraocular lenses (IOLs) was investigated in vitro. After two hours of lens contact with human granulocytes, scanning electron microscopy showed significantly less cell activation and granulocyte adhesion on the surface-modified IOL than on the untreated IOL (P < .01). The x-ray photo-electron spectroscopy analysis of the CF4 plasma surface modification demonstrated that new compounds containing fluorine were homogeneously grafted onto the PMMA lens surface, resulting in a marked increase of contact angle. These in vitro results must be confirmed by in vivo studies of CF4 plasma surface modification of IOLs.