Merck
Response of the human hepatic tissue cultures Hep-G2 and WRL-68 to cocaine.
Journal of pharmacological and toxicological methods (1995-04-01)
P M Falk, R T Sabater, D D Carballo
ZUSAMMENFASSUNG
The hydrolytic metabolism of cocaine into benzoylecgonine, ecgonine methyl ester, and ecgonine was studied in the human hepatoma cell line Hep-G2 and in the nontumorigenic fetal hepatic cell line WRL-68. Also, the toxicological response of these cells to cocaine was compared to previously published results obtained with perfused liver cells and in vivo systems. Our experiments indicated that Hep-G2 appear to have similar metabolic and toxicological patterns to in vivo and perfused cell systems. The WRL-68 tissue culture system was found to be less similar. These results suggest Hep-G2 cells can be utilized to study cocaine metabolism and toxicology, and possibly in studies involving other xenobiotic compounds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Ecgonin -hydrochlorid -Lösung, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®