Merck
Neurobehavioral toxicity of triethyltin in rats as a function of age at postnatal exposure.
Neurotoxicology (1984-01-01)
P H Ruppert, K F Dean, L W Reiter
ZUSAMMENFASSUNG
Triethyltin (TET) has been shown to be neurotoxic when injected on postnatal day (PND) 5. In the present experiment we examined the toxicity of a single exposure to TET at several postnatal ages. Rat pups were injected ip with 0 (saline), 1.5, 3.0, or 6.0 mg/kg TET bromide on PND 1, 5, 10 or 15. In agreement with our previous data, PND-5 exposure to 6 mg/kg TET produced behavioral toxicity and decreased adult brain weight. High dose pups were less successful in descending on a rope at 20 and 21 days of age, and were hyperactive in figure-eight mazes at 29-30 and 57-58 days of age. The spatial distribution of activity was also altered: photocell counts were increased primarily in the figure-eight area of the maze. The size of the milk bands was reduced in 6 mg/kg pups injected on either PND 1 or PND 5. Preweaning growth was decreased following all injection ages; this reduction was most pronounced for pups exposed to TET on PND 1 and PND 5. Mating behavior was disrupted in 6 mg/kg males irrespective of age at exposure. These data demonstrate a differential sensitivity to the toxicity of TET during postnatal life, with maximal susceptibility on PND 5.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triethylzinnbromid, 97%