Merck
  • Hemoglobin S antigelation agents based on 5-bromotryptophan with potential for sickle cell anemia.
Hemoglobin S antigelation agents based on 5-bromotryptophan with potential for sickle cell anemia.
Journal of medicinal chemistry (1990-12-01)
P Z De Croos, P Sangdee, B L Stockwell, L Kar, E B Thompson, M E Johnson, B L Currie
ZUSAMMENFASSUNG
5-Bromotryptophan (5-BrTrp) is the most potent amino acid derivative reported in the literature to inhibit the gelation of hemoglobin S (from sickle cell anemia patients). Trp-Trp is also more potent than Trp as an antigelation agent. Therefore, we have prepared a series of dipeptides containing 5-BrTrp and evaluated the antigelation activity. 5-BrTrp-5-BrTrp is the most potent, i.e., 5.9 times the activity of Trp, followed by 5-BrTrp-Trp and then Trp-5-BrTrp. This improved antigelation potency for 5-BrTrp-5-BrTrp and 5-BrTrp-Trp is very significant and will be pursued further as lead compounds with potential for sickle cell anemia.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5-Brom-DL-Tryptophan, 99%