Merck
  • Wedelolactone inhibits adipogenesis through the ERK pathway in human adipose tissue-derived mesenchymal stem cells.
Wedelolactone inhibits adipogenesis through the ERK pathway in human adipose tissue-derived mesenchymal stem cells.
Journal of cellular biochemistry (2012-06-09)
Seyoung Lim, Hyun-Jun Jang, Eun Hee Park, Jung Kuk Kim, Jung-Min Kim, Eung-Kyun Kim, Kyungmoo Yea, Yun-Hee Kim, Whaseon Lee-Kwon, Sung Ho Ryu, Pann-Ghill Suh
ZUSAMMENFASSUNG
Wedelolactone is an herbal medicine that is used to treat septic shock, hepatitis and venom poisoning. Although in differentiated and cancer cells, wedelolactone has been identified as anti-inflammatory, growth inhibitory, and pro-apoptotic, the effects of wedelolactone on stem cell differentiation remain largely unknown. Here, we report that wedelolactone inhibits the adipogenic differentiation of human adipose tissue-derived mesenchymal stem cells (hAMSCs). Wedelolactone reduced the formation of lipid droplets and the expression of adipogenesis-related proteins, such as CCAAT enhancer-binding protein-α (C/EBP-α), peroxisome proliferator-activated receptor-γ (PPAR-γ), lipoprotein lipase (LPL), and adipocyte fatty acid-binding protein aP2 (aP2). Wedelolactone mediated this process by sustaining ERK activity. In addition, inhibition of ERK activity with PD98059 resulted in reversion of the wedelolactone-mediated inhibition of adipogenic differentiation. Taken together, these results indicate that wedelolactone inhibits adipogenic differentiation through ERK pathway and suggest a novel inhibitory effect of wedelolactone on adipogenic differentiation in hAMSCs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wedelolacton, ≥98% (HPLC), powder