Merck
  • Bisphenol s, a new bisphenol analogue, in paper products and currency bills and its association with bisphenol a residues.
Bisphenol s, a new bisphenol analogue, in paper products and currency bills and its association with bisphenol a residues.
Environmental science & technology (2012-05-18)
Chunyang Liao, Fang Liu, Kurunthachalam Kannan
ZUSAMMENFASSUNG
As the evidence of the toxic effects of bisphenol A (BPA) grows, its application in commercial products is gradually being replaced with other related compounds, such as bisphenol S (BPS). Nevertheless, very little is known about the occurrence of BPS in the environment. In this study, BPS was analyzed in 16 types of paper and paper products (n = 268), including thermal receipts, paper currencies, flyers, magazines, newspapers, food contact papers, airplane luggage tags, printing paper, kitchen rolls (i.e., paper towels), and toilet paper. All thermal receipt paper samples (n = 111) contained BPS at concentrations ranging from 0.0000138 to 22.0 mg/g (geometric mean: 0.181 mg/g). The overall mean concentrations of BPS in thermal receipt papers were similar to the concentrations reported earlier for BPA in the same set of samples. A significant negative correlation existed between BPS and BPA concentrations in thermal receipt paper samples (r = -0.55, p < 0.0001). BPS was detected in 87% of currency bill samples (n = 52) from 21 countries, at concentrations ranging from below the limit of quantification (LOQ) to 6.26 μg/g (geometric mean: 0.029 μg/g). BPS also was found in 14 other paper product types (n = 105), at concentrations ranging from <LOQ to 8.38 μg/g (geometric mean: 0.0036 μg/g; detection rate: 52%). The estimated daily intake (EDI) of BPS, through dermal absorption via handling of papers and currency bills, was estimated on the basis of concentrations and frequencies of the handling of papers by humans. The median and 95th percentile EDI values, respectively, were 4.18 and 11.0 ng/kg body weight (bw)/day for the general population and 312 and 821 ng/kg bw/day for occupationally exposed individuals. Among the paper types analyzed, thermal receipt papers were found to be the major sources of human exposure to BPS (>88%). To our knowledge, this is the first report on the occurrence of BPS in paper products and currency bills.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4,4′-Sulfonyldiphenol, 98%